Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026647.3 | NP_080923.2 | 9 | 4356 | ||
GenBank mRNA | AK017682.1 | - | 1 | 2716 | ||
BC043110.1 | - | 9 | 4203 | 6406930 |