Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026908.3 | NP_081184.3 | 11 | 2098 | ||
GenBank mRNA | AK030474.1 | - | 11 | 2112 | ||
AK053642.1 | - | 11 | 2099 |