Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Eci3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026947.4 | NP_081223.1 | 5 | 493 | ||
GenBank mRNA | AK007578.1 | - | 5 | 494 | ||
AK131590.1 | - | 5 | 494 | |||
BC014724.1 | - | 5 | 467 | 4219360 | ||
FJ687155.1 | - | 4 | 416 |