Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026947.4 | NP_081223.1 | 8, 9 | 865 | ||
GenBank mRNA | AK007578.1 | - | 8, 9 | 866 | ||
AK131590.1 | - | 8, 9 | 866 | |||
BC014724.1 | - | 8, 9 | 839 | 4219360 | ||
FJ687155.1 | - | 7, 8 | 788 |