Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cnot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026949.3 | NP_081225.1 | 3 | 325 | ||
GenBank mRNA | AK007581.1 | - | 3 | 389 | ||
AK075727.1 | - | 3 | 295 | |||
AK150757.1 | - | 3 | 406 | |||
AK153907.1 | - | 3 | 328 | |||
AY515608.1 | - | 2 | 292 | |||
BC004040.1 | - | 3 | 325 | 3496188 |