Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027008.2 | NP_081284.2 | 2 | 366 | ||
GenBank mRNA | AK011637.1 | - | 2 | 101 | ||
AK052960.1 | - | 2 | 368 | |||
AK129077.1 | - | 2 | 326 | |||
AK143644.1 | - | 2 | 334 | |||
AK187166.1 | - | 2 | 83 | |||
BC021777.1 | - | 2 | 318 | 4215964 |