Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110209.1 | NP_001103679.1 | 4 | 414 | ||
NM_027133.3 | NP_081409.1 | 4 | 414 | |||
GenBank mRNA | AK009311.1 | - | 4 | 388 | ||
AK014897.1 | - | 4 | 312 | |||
AK053593.1 | - | 4 | 383 | |||
AK082900.1 | - | 4 | 398 | |||
AK136506.1 | - | 4 | 389 | |||
AK137445.1 | - | 4 | 415 | |||
AK143497.1 | - | 4 | 383 | |||
AK173241.1 | - | 4 | 346 | |||
AY312281.1 | - | 4 | 335 | |||
BC057961.1 | - | 4 | 341 | 5353629 | ||
BC060153.1 | - | 3 | 253 |