Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc52a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164819.1 | NP_001158291.1 | 5 | 2485 | ||
NM_001164820.1 | NP_001158292.1 | 4 | 1997 | |||
NM_027172.3 | NP_081448.2 | 5 | 2553 | |||
GenBank mRNA | AK009850.1 | - | 5 | 2436 | ||
AK047618.1 | - | 5 | 2485 | |||
AK154970.1 | - | 5 | 2535 | |||
BC016127.1 | - | 4 | 1966 | 4910227 | ||
CF424499.1 | - | 1 | 250 |
Set Membership: |
![]() |