Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027175.3 | NP_081451.3 | 3 | 874 | ||
GenBank mRNA | AK010328.1 | - | 2 | 704 | ||
AK050343.1 | - | 3 | 875 | |||
BC018422.1 | - | 3 | 704 | 3593650 |