Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027193.2 | NP_081469.2 | 8 | 1518 | ||
GenBank mRNA | BC060372.1 | - | 8 | 1518 | 6391546 |