Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164336.1 | NP_001157808.1 | 5 | 922 | ||
NM_027314.3 | NP_081590.3 | 5 | 922 | |||
GenBank mRNA | AK007511.1 | - | 5 | 758 | ||
AK009232.1 | - | 5 | 908 | |||
AK009364.1 | - | 5 | 920 | |||
AK178252.1 | - | 4 | 429 | |||
AK178944.1 | - | 4 | 436 | |||
AK188043.1 | - | 2 | 226 | |||
AK206577.1 | - | 3 | 317 | |||
AK219813.1 | - | 4 | 384 | |||
BC064752.1 | - | 5 | 646 | 30049608 | ||
BC107214.1 | - | 5 | 629 | |||
BC107215.1 | - | 5 | 634 | 40051628 | ||
BC132455.1 | - | 5 | 686 | 40130732 | ||
BC132457.1 | - | 5 | 685 | 40130734 |