Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159714.1 | NP_001153186.1 | 14 | 1882 | ||
NM_027328.4 | NP_081604.3 | 14 | 1900 | |||
GenBank mRNA | AK033190.1 | - | 14 | 1903 | ||
AK033283.1 | - | 14 | 1865 | |||
AK051260.1 | - | 14 | 1860 |