Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097590.2 | NP_001091059.1 | 3 | 640 | ||
NM_001265601.1 | NP_001252530.1 | 2 | 878 | |||
NM_006414.4 | NP_006405.2 | 2 | 521 | |||
NM_183005.4 | NP_892117.1 | 3 | 997 | |||
XM_006717363.1 | XP_006717426.1 | 3 | 811 | |||
XM_006717364.3 | XP_006717427.1 | 2 | 433 | |||
XM_011519293.1 | XP_011517595.1 | 2 | 676 | |||
XM_017015479.1 | XP_016870968.1 | 3 | 1129 | |||
XM_017015480.1 | XP_016870969.1 | 3 | 587 | |||
XM_017015481.1 | XP_016870970.1 | 2 | 611 | |||
GenBank mRNA | AK057006.1 | - | 3 | 795 | ||
AK223418.1 | - | 2 | 498 | |||
BC029494.1 | - | 3 | 778 | 5271044 | ||
BP247818.1 | - | 2 | 518 | |||
BU180901.1 | - | 2 | 638 | |||
CR541816.1 | - | 1 | 281 | |||
CX872398.1 | - | 3 | 572 | |||
U77664.1 | - | 2 | 317 |