Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027727.2 | NP_082003.1 | 3 | 425 | ||
GenBank mRNA | AK016957.1 | - | 3 | 354 | ||
AK049909.1 | - | 3 | 390 | |||
AK080149.1 | - | 3 | 402 | |||
AK081242.1 | - | 3 | 417 | |||
AK149628.1 | - | 3 | 399 | |||
AK149879.1 | - | 3 | 400 | |||
AK149891.1 | - | 3 | 399 | |||
AK154794.1 | - | 3 | 414 | |||
AK199364.1 | - | 1 | 36 | |||
BC038276.1 | - | 3 | 427 | 4458792 | ||
BC092283.1 | - | 3 | 411 |