Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285917.1 | NP_001272846.1 | 4 | 624 | ||
NM_029653.3 | NP_083929.2 | 4 | 624 | |||
NM_134062.2 | NP_598823.1 | 4 | 509 | |||
GenBank mRNA | AK009701.1 | - | 4 | 514 | ||
AK145922.1 | - | 4 | 510 | |||
AY245540.2 | - | 4 | 617 | |||
AY245541.1 | - | 4 | 610 | |||
BC021490.1 | - | 2 | 108 | |||
BC057317.1 | - | 4 | 529 | 6821014 | ||
BC060161.1 | - | 4 | 529 | 6821014 | ||
X97048.1 | - | 3 | 459 |
Set Membership: |
![]() |