Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030566.2 | NP_085043.2 | 13 | 2445 | ||
GenBank mRNA | AK029188.1 | - | 10 | 1747 | ||
AK089938.1 | - | 13 | 2151 | |||
AK134248.1 | - | 654 | ||||
AK144836.1 | - | 13 | 2131 | |||
AK157832.1 | - | 13 | 2106 | |||
BC004088.1 | - | 12 | 2191 | 3593722 | ||
BC015287.1 | - | 12 | 2694 | 4240442 | ||
BY665800.1 | - | 1 | 293 |