Gene Symbol: Smim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133697.3 | NP_598458.1 | 5 | 459 | ||
GenBank mRNA | AK003345.1 | - | 5 | 454 | ||
AK018994.1 | - | 5 | 468 | |||
AK044669.1 | - | 5 | 563 | |||
AK088833.1 | - | 5 | 506 | |||
AK164394.1 | - | 5 | 460 | |||
BC009097.1 | - | 5 | 454 | 3481841 | ||
BC029314.1 | - | 2 | 95 |