Search Thermo Fisher Scientific
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133701.2 | NP_598462.1 | 8 | 1030 | ||
GenBank mRNA | AK081998.1 | - | 8 | 965 | ||
AK145461.1 | - | 8 | 1033 | |||
AK166890.1 | - | 8 | 982 | |||
AK178683.1 | - | 1 | 34 | |||
AK189258.1 | - | 1 | 34 | |||
BC014869.1 | - | 8 | 957 | 3602158 | ||
BC023691.1 | - | 8 | 959 | 5344332 | ||
BC025030.1 | - | 8 | 976 | 4947416 |