Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133786.3 | NP_598547.1 | 6 | 922 | ||
GenBank mRNA | AJ534940.2 | - | 5 | 798 | ||
AK010848.1 | - | 4 | 373 | |||
AK048650.1 | - | 1 | 134 | |||
AK088350.1 | - | 6 | 905 | |||
AK088846.1 | - | 6 | 888 | |||
AK133817.1 | - | 6 | 912 | |||
AK152804.1 | - | 6 | 949 | |||
BC061481.1 | - | 6 | 897 | 6512832 | ||
BC062939.1 | - | 6 | 922 | 6841276 |