Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133831.3 | NP_598592.2 | 11 | 1384 | ||
GenBank mRNA | AK077341.1 | - | 11 | 1338 | ||
AK079167.1 | - | 11 | 1338 | |||
AK088461.1 | - | 11 | 1341 | |||
AK140286.1 | - | 209 | ||||
AK146916.1 | - | 11 | 1353 | |||
AK151836.1 | - | 11 | 1341 | |||
AK158665.1 | - | 11 | 1284 | |||
AK160070.1 | - | 11 | 1340 | |||
AK169290.1 | - | 11 | 1344 | |||
AY535001.1 | - | 11 | 1329 | |||
BC017637.1 | - | 11 | 1360 | 4912415 | ||
BC025810.1 | - | 11 | 1291 |