Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144525.3 | NP_653108.2 | 2 | 199 | ||
GenBank mRNA | AK034883.1 | - | 2 | 201 | ||
AK053521.1 | - | 215 | ||||
AK143078.1 | - | 201 | ||||
AK165285.1 | - | 2 | 211 | |||
AK166510.1 | - | 2 | 194 | |||
BC026651.1 | - | 2 | 115 | 4987771 | ||
BC029150.1 | - | 2 | 187 | 4954376 |