Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144525.3 | NP_653108.2 | 9 | 1120 | ||
GenBank mRNA | AK034883.1 | - | 9 | 1122 | ||
AK053521.1 | - | 1410 | ||||
AK143078.1 | - | 1122 | ||||
AK165285.1 | - | 8 | 1283 | |||
AK166510.1 | - | 9 | 1115 | |||
AK180903.1 | - | 1 | 27 | |||
BC022142.1 | - | 5 | 397 | 5101205 | ||
BC022602.1 | - | 3 | 266 | 4161204 | ||
BC026651.1 | - | 9 | 1036 | 4987771 | ||
BC027046.1 | - | 3 | 414 | 5362953 | ||
BC029150.1 | - | 9 | 1108 | 4954376 |