Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006453.2 | NP_006444.2 | 11 | 1118 | ||
GenBank mRNA | AB209173.1 | - | 11 | 1090 | ||
AK095076.1 | - | 6 | 593 | |||
BC007620.1 | - | 3 | 302 | |||
BC010231.1 | - | 11 | 1118 | 3455740 | ||
BC014007.2 | - | 11 | 1090 | 4040312 | ||
BC035409.1 | - | 11 | 1074 | 5165833 | ||
U02609.1 | - | 11 | 1066 |
Set Membership: |
![]() |