Gene Symbol: Dnajc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172400.3 | NP_765988.2 | 3, 4 | 262 | ||
GenBank mRNA | AK003892.2 | - | 3, 4 | 252 | ||
AK049526.1 | - | 3, 4 | 255 | |||
AK088325.1 | - | 3, 4 | 266 | |||
AK150933.1 | - | 3, 4 | 257 | |||
AK168645.1 | - | 3, 4 | 246 | |||
BC035539.1 | - | 3, 4 | 221 | |||
BC057191.1 | - | 3, 4 | 233 | |||
BC064732.1 | - | 3, 4 | 241 | |||
BC066222.1 | - | 3, 4 | 241 | 30459247 | ||
BC080760.1 | - | 3, 4 | 241 | |||
BC137735.1 | - | 3, 4 | 258 | 8860754 | ||
BC145655.1 | - | 3, 4 | 258 | |||
BY754536.1 | - | 3, 4 | 266 |