Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175145.3 | NP_780354.1 | 4 | 1660 | ||
GenBank mRNA | AK030324.1 | - | 4 | 1681 | ||
AK050216.1 | - | 4 | 1630 | |||
AK165880.1 | - | 4 | 1728 | |||
BC022794.1 | - | 1 | 799 | |||
BC049832.1 | - | 4 | 1660 | 6313802 |