Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164578.1 | NP_001158050.1 | 3 | 255 | ||
NM_175146.4 | NP_780355.2 | 3 | 255 | |||
GenBank mRNA | AK044504.1 | - | 3 | 231 | ||
BC025008.1 | - | 3 | 218 | |||
BC037444.1 | - | 2 | 119 | |||
BC049668.1 | - | 3 | 237 | 6535058 | ||
BG083382.2 | - | 2 | 128 | |||
CF732210.1 | - | 3 | 219 | |||
CJ072126.1 | - | 3 | 257 |