Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175155.4 | NP_780364.3 | 20 | 4491 | ||
GenBank mRNA | AJ507736.1 | - | 20 | 4306 | ||
AK039638.1 | - | 8 | 2830 | |||
AK129216.1 | - | 15 | 3825 | |||
AK197316.1 | - | 1 | 259 |