Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178027.4 | NP_821170.1 | 2 | 823 | ||
GenBank mRNA | AK031589.1 | - | 2 | 826 | ||
AK035321.1 | - | 2 | 776 | |||
AK150929.1 | - | 2 | 744 | |||
AK155578.1 | - | 2 | 784 | |||
AK165539.1 | - | 2 | 762 | |||
AK171154.1 | - | 2 | 782 | |||
AK185841.1 | - | 2 | 277 | |||
AK192911.1 | - | 2 | 280 | |||
AK194762.1 | - | 2 | 277 | |||
AK214365.1 | - | 2 | 273 | |||
BC049180.1 | - | 2 | 786 | 6314347 | ||
BC052721.1 | - | 2 | 747 | 6809853 | ||
BC062972.1 | - | 2 | 747 | 6809878 |