Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810026J23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178619.4 | NP_848734.1 | 2 | 2583 | ||
GenBank mRNA | AK007608.1 | - | 1 | 481 | ||
AK028799.1 | - | 2 | 2414 | |||
AK034762.1 | - | 2 | 2279 | |||
AK088045.1 | - | 2 | 2356 | |||
AK089121.1 | - | 2 | 2342 | |||
AK211362.1 | - | 1 | 87 | |||
AK213438.1 | - | 1 | 86 | |||
BC025883.1 | - | 2 | 2341 | |||
BC057071.1 | - | 2 | 2583 | 5695590 |