Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197992.1 | NP_932109.1 | 4 | 368 | ||
GenBank mRNA | AK008048.1 | - | 4 | 350 | ||
BC028560.1 | - | 4 | 368 | 1380201 |