Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020520.4 | NP_065266.1 | 9 | 1612 | ||
GenBank mRNA | AK075624.1 | - | 9 | 1613 | ||
AK152436.1 | - | 9 | 1583 | |||
BC029733.1 | - | 9 | 1610 | 4019406 | ||
BC052871.1 | - | 9 | 1535 | 30032324 |