Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srd5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020611.4 | NP_065636.2 | 4 | 568 | ||
GenBank mRNA | AK040198.1 | - | 4 | 536 | ||
AK075635.1 | - | 4 | 564 | |||
AK085748.1 | - | 4 | 417 | |||
AK138681.1 | - | 4 | 560 | |||
AK139290.1 | - | 4 | 565 | |||
AK162527.1 | - | 4 | 536 | |||
BC039564.1 | - | 4 | 521 |