Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290994.1 | NP_001277923.1 | 9 | 2805 | ||
NM_021323.3 | NP_067298.2 | 8 | 2741 | |||
GenBank mRNA | AF229257.1 | - | 9 | 2805 | ||
EU233992.1 | - | 5 | 2082 |