Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135727.2 | NP_001129199.1 | 18 | 3286 | ||
NM_001135728.2 | NP_001129200.1 | 13 | 2288 | |||
NM_001290661.1 | NP_001277590.1 | 7 | 1968 | |||
NM_001290664.1 | NP_001277593.1 | 6 | 1839 | |||
NM_021389.6 | NP_067364.2 | 17 | 2851 | |||
GenBank mRNA | AK004636.1 | - | 17 | 2851 | ||
AK028299.1 | - | 13 | 2292 | |||
AK049182.1 | - | 18 | 3288 | |||
AK052260.1 | - | 6 | 1514 | |||
AK080930.1 | - | 18 | 3152 | |||
AK135598.1 | - | 682 | ||||
AK142295.1 | - | 7 | 1742 | |||
AK161735.1 | - | 13 | 2369 | |||
AK172022.1 | - | 18 | 3267 | |||
AK200438.1 | - | 1 | 80 | |||
BC034646.1 | - | 17 | 2859 | |||
BC052863.1 | - | 17 | 2796 |