Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mmp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164197.1 | NP_001157669.1 | 5 | 577 | ||
NM_021412.2 | NP_067387.1 | 5 | 712 | |||
GenBank mRNA | AF153199.1 | - | 5 | 664 | ||
AF155221.1 | - | 5 | 692 | |||
AF162446.1 | - | 5 | 692 | |||
AK079924.1 | - | 5 | 578 | |||
AK080125.1 | - | 5 | 572 | |||
AK086808.1 | - | 4 | 452 | |||
AK136517.1 | - | 5 | 566 | |||
AK154863.1 | - | 5 | 706 | |||
BC112966.2 | - | 5 | 684 | 40039023 |