Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nudt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021431.2 | NP_067406.2 | 2 | 985 | ||
GenBank mRNA | AB041576.1 | - | 2 | 731 | ||
AK036077.1 | - | 2 | 316 | |||
AK048263.1 | - | 2 | 988 | |||
AK053310.1 | - | 2 | 765 | |||
AY152853.1 | - | 2 | 731 | |||
BC071274.1 | - | 2 | 789 | 6331160 |