Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021434.5 | NP_067409.2 | 9 | 1986 | ||
GenBank mRNA | AK053422.1 | - | 9 | 1956 |