Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021491.3 | NP_067466.1 | 3 | 922 | ||
GenBank mRNA | AJ250461.1 | - | 1 | 453 | ||
AK019476.2 | - | 4 | 974 | |||
AK076416.1 | - | 3 | 899 | |||
AK139226.1 | - | 3 | 898 | |||
BC043077.1 | - | 3 | 922 | 5721118 | ||
BC046980.1 | - | 2 | 742 | 6399438 |