Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021500.2 | NP_067475.2 | 8 | 1048 | ||
GenBank mRNA | AF263247.1 | - | 8 | 1010 | ||
AK034859.1 | - | 8 | 1047 | |||
AK049424.1 | - | 8 | 1068 | |||
AK075637.1 | - | 7 | 888 | |||
AK088854.1 | - | 8 | 1056 | |||
AK145800.1 | - | 8 | 1111 | |||
AK152523.1 | - | 8 | 1056 | |||
AK168788.1 | - | 8 | 1074 | |||
AK169149.1 | - | 8 | 1056 | |||
BC039054.1 | - | 8 | 1034 | 2648667 | ||
BC058687.1 | - | 8 | 1046 | 6411707 |