Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146001.1 | NP_001139473.1 | 9 | 1750 | ||
NM_021517.2 | NP_067492.2 | 9 | 1796 | |||
GenBank mRNA | AF220100.1 | - | 9 | 1735 | ||
AF474247.1 | - | 8 | 1734 | |||
AK006269.1 | - | 9 | 2270 | |||
AK012070.1 | - | 9 | 1786 | |||
AK014879.1 | - | 9 | 2053 | |||
AK029764.1 | - | 9 | 2263 | |||
AK165415.1 | - | 9 | 1733 | |||
BC013512.1 | - | 8 | 1719 | 4234949 |