Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021552.5 | NP_067527.3 | 5 | 745 | ||
GenBank mRNA | AF238866.1 | - | 5 | 671 | ||
AK002474.1 | - | 4 | 596 | |||
AK009136.1 | - | 4 | 674 | |||
AK012832.1 | - | 5 | 686 | |||
AK014275.1 | - | 5 | 722 | |||
AK017605.1 | - | 5 | 686 | |||
AK078457.1 | - | 5 | 698 | |||
BC086776.1 | - | 4 | 541 | |||
BC108365.1 | - | 5 | 606 | 4219428 | ||
BC116384.1 | - | 5 | 686 | |||
BC126976.1 | - | 5 | 685 | 40101726 | ||
BC172160.1 | - | 5 | 657 |