Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021554.2 | NP_067529.2 | 1, 2 | 527 | ||
GenBank mRNA | AB041664.1 | - | 1, 2 | 305 | ||
AJ278576.1 | - | 1, 2 | 527 | |||
AK075586.1 | - | 1, 2 | 290 | |||
AK201397.1 | - | 1, 2 | 15 | |||
BC068124.1 | - | 1, 2 | 321 | 5716972 |