Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021559.2 | NP_067534.2 | 4 | 2008 | ||
GenBank mRNA | AK076173.1 | - | 1 | 731 | ||
AK088914.1 | - | 1 | 1041 | |||
AK146726.1 | - | 4 | 2006 | |||
AY832930.1 | - | 4 | 2047 | |||
BC054832.1 | - | 4 | 2008 | 6467818 |