Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006613.3 | NP_006604.1 | 2 | 198 | ||
XM_017024029.1 | XP_016879518.1 | 3 | 330 | |||
XM_017024030.1 | XP_016879519.1 | 3 | 333 | |||
GenBank mRNA | BC063035.1 | - | 2 | 170 | 5757176 | |
BT019981.1 | - | 2 | 101 | |||
CR541941.1 | - | 2 | 101 | |||
CR541966.1 | - | 2 | 101 | |||
U52518.1 | - | 2 | 134 |
Gene Symbol: GRAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129778.1 | NP_001123250.1 | 2 | 252 | ||
XM_011523857.2 | XP_011522159.1 | 3 | 328 | |||
XM_017024635.1 | XP_016880124.1 | 3 | 335 | |||
XM_017024636.1 | XP_016880125.1 | 2 | 314 | |||
XM_017024637.1 | XP_016880126.1 | 3 | 331 | |||
GenBank mRNA | BC157869.1 | - | 2 | 252 | 9057088 |