Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021899.3 | NP_068699.1 | 7 | 1653 | ||
GenBank mRNA | AB221666.1 | - | 6 | 852 | ||
AF253052.1 | - | 7 | 1665 | |||
BC040395.1 | - | 8 | 1283 | 4190087 |