Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022018.3 | NP_071301.2 | 11, 12 | 1608 | ||
GenBank mRNA | AB049355.1 | - | 11, 12 | 1448 | ||
AK029847.1 | - | 7, 8 | 920 | |||
AK136597.1 | - | 11, 12 | 1518 | |||
AK170193.1 | - | 11, 12 | 1531 | |||
AK170347.1 | - | 2, 3 | 203 | |||
AK210084.1 | - | 1, 2 | 97 | |||
BC021332.1 | - | 11, 12 | 1496 | |||
BC038006.1 | - | 11, 12 | 1464 | |||
BC058234.1 | - | 9, 10 | 1208 | 6306849 | ||
BC099690.1 | - | 11, 12 | 1515 | |||
BC137843.1 | - | 7, 8 | 985 | 8860862 |