Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022328.2 | NP_071723.1 | 12 | 3505 | ||
GenBank mRNA | AK156820.1 | - | 12 | 3532 | ||
BC053005.1 | - | 12 | 3505 | 5716396 | ||
BC139810.1 | - | 3 | 2013 |