Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022413.4 | NP_071858.3 | 4 | 684 | ||
GenBank mRNA | AB037373.1 | - | 4 | 585 | ||
AJ542487.1 | - | 4 | 389 | |||
AK077234.1 | - | 4 | 614 | |||
AK085123.1 | - | 3 | 997 | |||
AK168760.1 | - | 4 | 684 | |||
BC016101.1 | - | 4 | 622 | 4911355 |