Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gpr88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022427.2 | NP_071872.1 | 2 | 2135 | ||
GenBank mRNA | AB042408.1 | - | 2 | 2105 | ||
BC051941.1 | - | 1 | 1751 | 5708059 |
Set Membership: |
![]() |