Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022886.2 | NP_075024.2 | 5 | 402 | ||
GenBank mRNA | AF245700.2 | - | 5 | 402 | ||
AK020346.1 | - | 5 | 324 | |||
BC025142.1 | - | 5 | 380 | |||
BC111874.1 | - | 5 | 322 | 40059586 | ||
BC112380.1 | - | 5 | 323 | 40059585 |